Assay Details
| Gene Symbol: IER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075202.2 | NP_001068670.1 | 1 - 2 | 244 | 100 | |
| GenBank mRNA | BC120062.1 | - | 1 - 2 | 245 | 100 | |
| EV690072.1 | - | 1 - 2 | 245 | 100 | ||
More Information
Set Membership: |
|

IER3
Genomic Map