Assay Details
| Gene Symbol: S100A16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075218.1 | NP_001068686.1 | 1 - 2 | 127 | 138 | |
| GenBank mRNA | BC120103.1 | - | 1 - 2 | 127 | 138 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC120103.1, NM_001075218.1 transcripts.
Set Membership: |
|

S100A16
Genomic Map