Assay Details
Gene Symbol: GK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075236.1 | NP_001068704.1 | 8 - 9 | 885 | 64 | |
GenBank mRNA | BC122692.1 | - | 8 - 9 | 885 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
