Assay Details
| Gene Symbol: AAAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075269.1 | NP_001068737.1 | 2 - 3 | 323 | 63 | |
| GenBank mRNA | BC120418.1 | - | 2 - 3 | 323 | 63 | |
More Information
Set Membership: |
|

AAAS
Genomic Map