Assay Details
| Gene Symbol: PCBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075283.2 | NP_001068751.1 | 12 - 13 | 1079 | 59 | |
| GenBank mRNA | BC120009.1 | - | 12 - 13 | 1080 | 59 | |
More Information
Set Membership: |
|

PCBP4
Genomic Map