Assay Details
| Gene Symbol: WSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075357.1 | NP_001068825.1 | 3 - 4 | 617 | 64 | |
| GenBank mRNA | BC149531.1 | - | 3 - 4 | 571 | 64 | |
| BT026227.1 | - | 3 - 4 | 617 | 64 | ||
More Information
Set Membership: |
|

WSB2
Genomic Map