Assay Details
Gene Symbol: CDK5RAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075399.1 | NP_001068867.1 | 4 - 5 | 337 | 64 | |
GenBank mRNA | BC119981.1 | - | 4 - 5 | 337 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
