Assay Details
Gene Symbol: PSKH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075413.1 | NP_001068881.1 | 2 - 3 | 1134 | 76 | |
GenBank mRNA | BT026549.1 | - | 2 - 3 | 1134 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
