Assay Details
| Gene Symbol: IFIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075414.1 | NP_001068882.1 | 1 - 2 | 59 | 77 | |
| GenBank mRNA | BC123421.1 | - | 1 - 2 | 59 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC123421.1, NM_001075414.1 transcripts.
Set Membership: |
|

IFIT3
Genomic Map