Assay Details
| Gene Symbol: MRPL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075445.1 | NP_001068913.1 | 4 - 5 | 613 | 63 | |
| GenBank mRNA | BC120390.1 | - | 4 - 5 | 613 | 63 | |
More Information
Set Membership: |
|

MRPL15
Genomic Map