Assay Details
Gene Symbol: IFI35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075462.2 | NP_001068930.1 | 4 - 5 | 537 | 69 | |
GenBank mRNA | BC122808.1 | - | 4 - 5 | 538 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
