Assay Details
| Gene Symbol: CHRAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075478.1 | NP_001068946.1 | 2 - 3 | 271 | 113 | |
| GenBank mRNA | BC120142.1 | - | 2 - 3 | 271 | 113 | |
More Information
Set Membership: |
|

CHRAC1
Genomic Map