Assay Details
Gene Symbol: ST6GALNAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075533.1 | NP_001069001.1 | 7 - 8 | 922 | 71 | |
GenBank mRNA | BC118158.1 | - | 7 - 8 | 922 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
