Assay Details
| Gene Symbol: CES1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075554.1 | NP_001069022.1 | 1 - 2 | 116 | 67 | |
| GenBank mRNA | BC120153.1 | - | 1 - 2 | 116 | 67 | |
More Information
Set Membership: |
|

CES1
Genomic Map