Assay Details
| Gene Symbol: ADAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075577.1 | NP_001069045.1 | 6 - 7 | 661 | 63 | |
| GenBank mRNA | BC120402.1 | - | 6 - 7 | 661 | 63 | |
More Information
Set Membership: |
|

ADAL
Genomic Map