Assay Details
| Gene Symbol: RTF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075625.2 | NP_001069093.1 | 8 - 9 | 796 | 57 | |
| GenBank mRNA | BC120045.1 | - | 8 - 9 | 797 | 57 | |
More Information
Set Membership: |
|

RTF2
Genomic Map