Assay Details
| Gene Symbol: ALDH8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075626.1 | NP_001069094.1 | 2 - 3 | 310 | 56 | |
| GenBank mRNA | BC120092.1 | - | 2 - 3 | 310 | 56 | |
| BT029881.1 | - | 2 - 3 | 321 | 56 | ||
More Information
Set Membership: |
|

ALDH8A1
Genomic Map