Assay Details
Gene Symbol: SAP30BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075627.2 | NP_001069095.1 | 5 - 6 | 445 | 63 | |
GenBank mRNA | BC120252.1 | - | 5 - 6 | 448 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
