Assay Details
| Gene Symbol: RBM23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075636.1 | NP_001069104.1 | 2 - 3 | 255 | 77 | |
| GenBank mRNA | BC122593.1 | - | 2 - 3 | 255 | 77 | |
More Information
Set Membership: |
|

RBM23
Genomic Map