Assay Details
| Gene Symbol: ZPBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075661.2 | NP_001069129.1 | 3 - 4 | 386 | 86 | |
| GenBank mRNA | BC120176.1 | - | 3 - 4 | 387 | 86 | |
More Information
Set Membership: |
|

ZPBP2
Genomic Map