Assay Details
Gene Symbol: AP2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075702.1 | NP_001069170.1 | 3 - 4 | 402 | 61 | |
GenBank mRNA | BC120121.1 | - | 3 - 4 | 402 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
