Assay Details
| Gene Symbol: TOX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075707.1 | NP_001069175.1 | 6 - 7 | 905 | 71 | |
| GenBank mRNA | BC119929.1 | - | 6 - 7 | 905 | 71 | |
More Information
Set Membership: |
|

TOX4
Genomic Map