Assay Details
| Gene Symbol: MYL6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075713.1 | NP_001069181.1 | 3 - 4 | 363 | 74 | |
| GenBank mRNA | BC118334.1 | - | 3 - 4 | 363 | 74 | |
More Information
Set Membership: |
|

MYL6B
Genomic Map