Assay Details
Gene Symbol: CDK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075715.1 | NP_001069183.1 | 11 - 12 | 1100 | 121 | |
GenBank mRNA | BC123525.1 | - | 11 - 12 | 1100 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
