Assay Details
| Gene Symbol: VAPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075733.1 | NP_001069201.1 | 5 - 6 | 769 | 124 | |
| GenBank mRNA | BC119937.1 | - | 5 - 6 | 769 | 124 | |
More Information
Set Membership: |
|

VAPA
Genomic Map