Assay Details
| Gene Symbol: NAA50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075750.1 | NP_001069218.1 | 2 - 3 | 401 | 58 | |
| GenBank mRNA | BC122617.1 | - | 2 - 3 | 401 | 58 | |
More Information
Set Membership: |
|

NAA50
Genomic Map