Assay Details
Gene Symbol: OSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075776.1 | NP_001069244.1 | 1 - 2 | 507 | 70 | |
GenBank mRNA | BC120014.1 | - | 1 - 2 | 507 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
