Assay Details
| Gene Symbol: TFE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075811.2 | NP_001069279.2 | 8 - 9 | 1155 | 57 | |
| GenBank mRNA | BC122581.1 | - | 8 - 9 | 1155 | 57 | |
More Information
Set Membership: |
|

TFE3
Genomic Map