Assay Details
| Gene Symbol: PLA2G2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075820.2 | NP_001069288.1 | 4 - 5 | 366 | 125 | |
| GenBank mRNA | BC122706.1 | - | 4 - 5 | 367 | 125 | |
More Information
Set Membership: |
|

PLA2G2A
Genomic Map