Assay Details
Gene Symbol: NKIRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075835.1 | NP_001069303.1 | 1 - 2 | 50 | 58 | |
GenBank mRNA | BC120341.1 | - | 1 - 2 | 50 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC120341.1, NM_001075835.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
