Assay Details
Gene Symbol: NKIRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075835.1 | NP_001069303.1 | 3 - 4 | 401 | 125 | |
GenBank mRNA | BC120341.1 | - | 3 - 4 | 401 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
