Assay Details
Gene Symbol: GAL3ST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075840.1 | NP_001069308.1 | 2 - 3 | 443 | 78 | |
GenBank mRNA | BC120166.1 | - | 2 - 3 | 443 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
