Assay Details
| Gene Symbol: SERPINB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075859.2 | NP_001069327.1 | 4 - 5 | 437 | 76 | |
| GenBank mRNA | BC123622.1 | - | 4 - 5 | 438 | 76 | |
More Information
Set Membership: |
|

SERPINB9
Genomic Map