Assay Details
| Gene Symbol: ENOPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075875.1 | NP_001069343.1 | 3 - 4 | 596 | 61 | |
| GenBank mRNA | BC119871.1 | - | 3 - 4 | 596 | 61 | |
More Information
Set Membership: |
|

ENOPH1
Genomic Map