Assay Details
Gene Symbol: SPOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075936.1 | NP_001069404.1 | 10 - 11 | 1248 | 78 | |
GenBank mRNA | BC119970.1 | - | 10 - 11 | 1248 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
