Assay Details
| Gene Symbol: SNAP29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075959.1 | NP_001069427.1 | 4 - 5 | 693 | 77 | |
| GenBank mRNA | BC122755.1 | - | 4 - 5 | 693 | 77 | |
More Information
Set Membership: |
|

SNAP29
Genomic Map