Assay Details
| Gene Symbol: TTI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076042.1 | NP_001069510.1 | 6 - 7 | 3149 | 68 | |
| GenBank mRNA | BC126508.1 | - | 7 - 8 | 3161 | 68 | |
| BT026193.1 | - | 6 - 7 | 3149 | 68 | ||
More Information
Set Membership: |
|

TTI1
Genomic Map