Assay Details
| Gene Symbol: ABHD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076063.2 | NP_001069531.1 | 4 - 5 | 744 | 64 | |
| GenBank mRNA | BC120233.1 | - | 4 - 5 | 745 | 64 | |
| JX446395.1 | - | 4 - 5 | 657 | 64 | ||
| JX446396.1 | - | 4 - 5 | 657 | 64 | ||
| JX446397.1 | - | 4 - 5 | 657 | 64 | ||
| JX446398.1 | - | 2 - 3 | 201 | 64 | ||
| KF539976.1 | - | 4 - 5 | 657 | 64 | ||
More Information
Set Membership: |
|

ABHD5
Genomic Map