Assay Details
Gene Symbol: GLIPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076112.2 | NP_001069580.1 | 4 - 5 | 358 | 78 | |
GenBank mRNA | BC120159.1 | - | 4 - 5 | 360 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
