Assay Details
Gene Symbol: TMEM110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076117.1 | NP_001069585.1 | 4 - 5 | 541 | 82 | |
GenBank mRNA | BC120361.1 | - | 4 - 5 | 541 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
