Assay Details
| Gene Symbol: TRAPPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076120.2 | NP_001069588.1 | 4 - 5 | 522 | 108 | |
| GenBank mRNA | BC120158.1 | - | 4 - 5 | 523 | 108 | |
More Information
Set Membership: |
|

TRAPPC3
Genomic Map