Assay Details
| Gene Symbol: PDE12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076133.1 | NP_001069601.1 | 2 - 3 | 1460 | 124 | |
| GenBank mRNA | BC123772.1 | - | 2 - 3 | 1460 | 124 | |
More Information
Set Membership: |
|

PDE12
Genomic Map