Assay Details
| Gene Symbol: EIF3J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076143.1 | NP_001069611.1 | 6 - 7 | 595 | 85 | |
| GenBank mRNA | BC119989.1 | - | 6 - 7 | 595 | 85 | |
| BC149417.1 | - | 6 - 7 | 678 | 85 | ||
More Information
Set Membership: |
|

EIF3J
Genomic Map