Assay Details
| Gene Symbol: SPRY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076147.1 | NP_001069615.1 | 1 | 1447 | 105 | |
| GenBank mRNA | BC123435.1 | - | 1 | 1447 | 105 | |
More Information
Set Membership: |
|

SPRY2
Genomic Map