Assay Details
| Gene Symbol: CD180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076168.1 | NP_001069636.1 | 2 - 3 | 374 | 102 | |
| GenBank mRNA | BC148876.1 | - | 2 - 3 | 382 | 102 | |
| BT026253.1 | - | 2 - 3 | 374 | 102 | ||
More Information
Set Membership: |
|

CD180
Genomic Map