Assay Details
| Gene Symbol: RAB5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076186.1 | NP_001069654.1 | 2 - 3 | 376 | 77 | |
| GenBank mRNA | BC122651.1 | - | 2 - 3 | 376 | 77 | |
More Information
Set Membership: |
|

RAB5A
Genomic Map