Assay Details
Gene Symbol: ALG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076189.1 | NP_001069657.2 | 2 - 3 | 488 | 63 | |
GenBank mRNA | BC119908.1 | - | 2 - 3 | 488 | 63 | |
BT030571.1 | - | 2 - 3 | 448 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
