Assay Details
| Gene Symbol: ACHE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076220.1 | NP_001069688.1 | 2 - 3 | 1125 | 94 | |
| GenBank mRNA | BC123898.1 | - | 2 - 3 | 1125 | 94 | |
More Information
Set Membership: |
|

ACHE
Genomic Map