Assay Details
Gene Symbol: MAGOHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076252.1 | NP_001069720.1 | 3 - 4 | 326 | 121 | |
GenBank mRNA | BC120291.1 | - | 3 - 4 | 326 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
