Assay Details
Gene Symbol: TUBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076255.2 | NP_001069723.1 | 3 - 4 | 428 | 71 | |
GenBank mRNA | BC120225.1 | - | 3 - 4 | 429 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
